Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc perchlorate hexahydrate ≥99.995% (trace metals basis)

Supplier: BeanTown Chemical


214835-50G 214835-250G 214835-10G
BT214835-50GEA 238.77 USD InStock
BT214835-50G BT214835-250G BT214835-10G
Zinc perchlorate hexahydrate ≥99.995% (trace metals basis)
Zinc perchlorate hexahydrate
CAS: 10025-64-6; EC No: 237-122-0; MDL No: MFCD00150370; RTECS: SD0525000
UN No: UN1481; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: Zn(ClO4)2·6H2O; MW: 372.37
Melting Point: 106°
Density (g/mL): 2.250; Refractive Index: 1.50


Formula: Cl₂O₈Zn·6H₂O
MW: 372,38 g/mol
Melting Pt: 106 °C
Density: 2.25 g/cm³ (20 °C)
MDL Number: MFCD00150370
CAS Number: 10025-64-6
EINECS: 237-122-0
UN: 1481
ADR: 5.1,II
Order Now
Specification Test Results
Assay 99.995%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty