Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 4pm ET Saturday through 8pm Sunday.

Lithium aluminum hydride ≥97%, Powder

Supplier: BeanTown Chemical

Synonyms: Lithium tetrahydridoaluminateAluminium lithium hydride

214805-250G 214805-25G 214805-100G
BT214805-250GEA 220.66 USD InStock
BT214805-250G BT214805-25G BT214805-100G
Lithium aluminum hydride ≥97%, Powder
Lithium aluminum hydride
CAS: 16853-85-3; EC No: 240-877-9; MDL No: MFCD00011075; RTECS: BD0100000
UN No: UN1410; Haz Class: 4.3; Packing Group: I
Liquid; Linear Formula: LiAlH4 ; MW: 37.95
Melting Point: 125° (decomposes)
Air Sensitive, Moisture Sensitive


Formula: H₄AlLi
MW: 37,95 g/mol
Melting Pt: 125 °C
Density: 0.91 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1410
ADR: 4.3,I
Merck Index: 13,00348
Order Now
Specification Test Results
Assay 97%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty