Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus(III) chloride ≥98%

Supplier: BeanTown Chemical

Synonyms: Phosphorus trichloride

214795-50G 214795-250G
BT214795-50GEA 24.58 USD InStock
BT214795-50G BT214795-250G
Phosphorus(III) chloride ≥98%
Phosphorus (III) chloride
CAS: 7719-12-2; EC No: 231-749-3; MDL No: MFCD00011438
UN No: UN1809; Haz Class: 6.1 (8); Packing Group: I
Liquid; Linear Formula: PCl3; MW: 137.33
Melting Point: -112°; Boiling Point: 74-78°
Density (g/mL): 1.574; Refractive Index: 1.515
Light Sensitive, Moisture Sensitive


Formula: Cl₃P
MW: 137,33 g/mol
Boiling Pt: 76 °C (1013 hPa)
Melting Pt: -112 °C
Density: 1.57 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00011438
CAS Number: 7719-12-2
EINECS: 231-749-3
UN: 1809
ADR: 6.1,I
Merck Index: 12,07515
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty