Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Antimony ≥99.5% (trace metals basis), Powder -200 mesh

Supplier: BeanTown Chemical


214710-500G 214710-100G
BT214710-500GEA 76.58 USD InStock
BT214710-500G BT214710-100G
Antimony ≥99.5% (trace metals basis), Powder -200 mesh
Antimony
CAS: 7440-36-0; EC No: 231-146-5; MDL No: MFCD00134030; RTECS: CC4025000
UN No: UN2871; Haz Class: 6.1; Packing Group: III
Powder; Molecular Formula: Sb; MW: 121.76
Melting Point: 630°; Boiling Point: 1635°
Density (g/mL): 6.69


Formula: Sb
MW: 121,76 g/mol
Boiling Pt: 1637 °C (1013 hPa)
Melting Pt: 630 °C
Density: 6.684 g/cm³ (20 °C)
MDL Number: MFCD00134030
CAS Number: 7440-36-0
EINECS: 231-146-5
UN: 2871
ADR: 6.1,III
Merck Index: 12,00733
Order Now
Specification Test Results
Assay 99.5%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty