Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Diethyl acetylenedicarboxylate ≥96%

Supplier: BeanTown Chemical


214705-100G 214705-500G 214705-25G
BT214705-100GEA 209.49 USD InStock
BT214705-100G BT214705-500G BT214705-25G
Diethyl acetylenedicarboxylate ≥96%
Diethyl acetylenedicarboxylate
CAS: 762-21-0; EC No: 212-095-8; MDL No: MFCD00009186
UN No: UN1760; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: CH3CH2O2CC≡CCO2CH2CH3; Molecular Formula: C8H10O4 ; MW: 170.16
Boiling Point: 107-110°/11 mmHg; Flash point: 94°C (201°F)
Density (g/mL): 1.063; Refractive Index: 1.443


Formula: C₈H₁₀O₄
MW: 170,16 g/mol
MDL Number: MFCD00009186
CAS Number: 762-21-0
EINECS: 212-095-8
Order Now
Specification Test Results
Assay 96%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty