Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 4pm ET Saturday through 8pm Sunday.

Praseodymium ≥99.9% (trace rare earth metals basis), Powder -40 mesh

Supplier: BeanTown Chemical


Ampuled under argon

214670-5G
BT214670-5GEA 112.63 USD InStock
BT214670-5G
Praseodymium ≥99.9% (trace rare earth metals basis), Powder -40 mesh
Praseodymium
CAS: 7440-10-0; EC No: 231-120-3; MDL No: MFCD00011174
UN No: UN3089; Haz Class: 4.1; Packing Group: II
Powder; Molecular Formula: Pr; MW: 140.91
Melting Point: 931°; Boiling Point: 3520°
Density (g/mL): 6.71


Formula: Pr
MW: 140,91 g/mol
MDL Number: MFCD00011174
CAS Number: 7440-10-0
EINECS: 231-120-3
UN: 3178
ADR: 4.1,III
Order Now
Specification Test Results
Assay 99.9%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty