Calcium hydroxide ≥95%

Supplier: BeanTown Chemical

Synonyms: Slaked lime

214620-1KG 214620-250G 214620-5KG
BT214620-1KGEA 50.22 USD
BT214620-1KG BT214620-250G BT214620-5KG
Calcium hydroxide ≥95%
Calcium hydroxide

  • RTECS: EW2800000
  • Air Sensitive
  • Powder

Formula: Ca(OH)₂
MW: 74.09 g/mol
Density: 2.24 g/mL
MDL Number: MFCD00010901
CAS Number: 1305-62-0
EINECS: 215-137-3
UN: 3262
ADR: 8,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR