trans-Dichloroethylene ≥98% stabilized

Supplier: BeanTown Chemical

Synonyms: trans-Dichloroethene(E)-1,2-Dichloroethenetrans-1,2-Dichloroethylene(E)-1,2-Dichloroethylene

214615-100G 214615-25G
BT214615-100GEA 198.37 USD
BT214615-100G BT214615-25G
trans-Dichloroethylene ≥98% stabilized
trans-Dichloroethylene

  • RTECS: KV9400000
  • Refractive index: 1.446
  • Liquid

Formula: C₂H₂Cl₂
MW: 96.94 g/mol
Boiling Pt: 47…49 °C
Melting Pt: –50 °C
Density: 1.257 g/mL
Flash Pt: 2 °C (35 °F)
MDL Number: MFCD00062942
CAS Number: 156-60-5
EINECS: 205-860-2
UN: 1150
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR