Tetrahydrofuran-[D₈] ≥99.5% (Isotopic)
Supplier: BeanTown Chemical
Synonyms:
THF-d8
, THF-D8
, Octadeuterotetrahydrofuran
, Tetrahydrofuran-d8
, Deuterated Tetrahydrofuran
, Tetrahydrofuran-[²H₈]
214460-1G
BT214460-1GEA
72.21
USD
BT214460-1G
Tetrahydrofuran-[D₈] ≥99.5% (Isotopic)
Tetrahydrofuran-[D8]
- Refractive index: 1.403
- Liquid
Formula:
C₄D₈O MW: 80.04 g/mol Boiling Pt: 65…66 °C Melting Pt: –106 °C Density: 0.985 g/mL Flash Pt: –17 °C (1 °F) |
MDL Number:
MFCD00044238 CAS Number: 1693-74-9 EINECS: 216-898-4 UN: 2056 ADR: 3,II |
Specification Test Results
Assay | 99.5% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...