Sodium hydroxide ≥98%, pellets

Supplier: BeanTown Chemical

Synonyms: Sodium hydrateSoda lyeCaustic sodaLye

214430-500G
BT214430-500GEA 28.86 USD
BT214430-500G
Sodium hydroxide ≥98%, pellets
Sodium hydroxide

  • RTECS: WB4900000
  • Air Sensitive, Hygroscopic
  • Pellets

Formula: NaOH
MW: 40 g/mol
Boiling Pt: 1390 °C
Melting Pt: 318 °C
Density: 2.13 g/mL
MDL Number: MFCD00003548
CAS Number: 1310-73-2
EINECS: 215-185-5
UN: 1823
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR