Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Iodobutane ≥99% stabilized

Supplier: BeanTown Chemical

Synonyms: 1-Butyl iodide

214415-500G 214415-100G
BT214415-500GEA 103.14 USD InStock
BT214415-500G BT214415-100G
1-Iodobutane ≥99% stabilized
1-Iodobutane
CAS: 542-69-8; EC No: 208-824-4; MDL No: MFCD00001098; RTECS: EK4400000
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3(CH2)3I; Molecular Formula: C4H9I; MW: 184.02
Melting Point: -103°; Boiling Point: 130-131°; Flash point: 33°C (91°F)
Density (g/mL): 1.617; Refractive Index: 1.498
Light Sensitive


Formula: C₄H₉I
MW: 184,02 g/mol
Boiling Pt: 130 °C (1013 hPa)
Melting Pt: -103 °C
Density: 1.616 g/cm³ (20 °C)
Flash Pt: 36 °C
Storage Temperature: ambient
MDL Number: MFCD00001098
CAS Number: 542-69-8
EINECS: 208-824-4
UN: 1993
ADR: 3,III
Merck Index: 13,01572
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty