Hept-6-enoic acid ≥98%
Supplier: BeanTown Chemical
Synonyms:
6-Heptenoic acid
214395-1G
214395-5G
BT214395-1GEA
105.58
USD
BT214395-1G
BT214395-5G
Hept-6-enoic acid ≥98%
Hept-6-enoic acid
- Refractive index: 1.439
- Liquid
Formula:
C₇H₁₂O₂ MW: 128.17 g/mol Boiling Pt: 222…224 °C Melting Pt: –6 °C Density: 0.946 g/mL Flash Pt: >110 °C (>230 °F) |
MDL Number:
MFCD00014382 CAS Number: 1119-60-4 EINECS: 214-283-5 UN: 3265 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...