p-Toluenesulfonyl chloride ≥98%

Synonyms: 4-Toluenesulfonyl chlorideToluene-4-sulfonyl chlorideToluene-p-sulfonyl chlorideToluene-4-sulphonyl chlorideTosyl chloride4-Toluenesulphonyl chlorideToluene-p-sulphonyl chloride

214290-250G 214290-1KG
BT214290-250GEA 26.72 USD
BT214290-250G BT214290-1KG
p-Toluenesulfonyl chloride ≥98%
p-Toluenesulphonyl chloride

  • RTECS: DB8929000
  • Moisture Sensitive
  • Crystalline

Formula: C₇H₇ClO₂S
MW: 190.65 g/mol
Boiling Pt: 134 °C/10 mmHg
Melting Pt: 65…69 °C
Density: 1.4 g/mL
Flash Pt: 128 °C (262 °F)
MDL Number: MFCD00007450
CAS Number: 98-59-9
EINECS: 202-684-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR