Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Methylmaleimide ≥98%

Supplier: BeanTown Chemical


214245-5G 214245-25G
BT214245-5GEA 47.96 USD InStock
BT214245-5G BT214245-25G
N-Methylmaleimide ≥98%
N-Methylmaleimide
CAS: 930-88-1; EC No: 213-226-1; MDL No: MFCD00005508; RTECS: ON5600000
UN No: UN2928; Haz Class: 6.1 (8); Packing Group: II
Solid; Molecular Formula: C5H5NO2; MW: 111.10
Melting Point: 94-96°


Formula: C₅H₅NO₂
MW: 111.1 g/mol
Boiling Pt: 194 °C (1013 hPa)
Melting Pt: 70 to 75 °C
MDL Number: MFCD00005508
CAS Number: 930-88-1
EINECS: 213-226-1
UN: 2928
ADR: 6.1,II
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty