Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Triethyl orthoacetate ≥98%

Supplier: BeanTown Chemical


214240-100ML 214240-500ML 214240-2.5L
BT214240-100MLEA 32.89 USD InStock
BT214240-100ML BT214240-500ML BT214240-2.5L
Triethyl orthoacetate ≥98%
Triethyl orthoacetate
CAS: 78-39-7; EC No: 201-112-4; MDL No: MFCD00009223
UN No: UN1993; Haz Class: 3; Packing Group: III
Liquid; Linear Formula: CH3C(OCH2CH3)3; Molecular Formula: C8H18O3; MW: 162.23
Boiling Point: 142°; Flash point: 32°C (90°F)
Density (g/mL): 0.885; Refractive Index: 1.396
Moisture Sensitive


Formula: C₈H₁₈O₃
MW: 162,23 g/mol
Boiling Pt: 142 °C (1013 hPa)
Density: 0.8847 g/cm³ (20 °C)
Flash Pt: 32 °C
MDL Number: MFCD00009223
CAS Number: 78-39-7
EINECS: 201-112-4
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty