Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cyclohexane ≥99.7% Environmental

Supplier: BeanTown Chemical

Danger
Synonyms: Hexahydro-BenzeneHexamethylene

214185-4X4L 214185-4L
BT214185-4X4LEA 390.01 USD InStock
BT214185-4X4L BT214185-4L
Cyclohexane ≥99.7% Environmental
Cyclohexane
CAS: 110-82-7; EC No: 203-806-2; MDL No: MFCD00003814; RTECS: GU6300000
UN No: UN1145; Haz Class: 3; Packing Group: II
Liquid; Molecular Formula: C6H12; MW: 84.16
Melting Point: 4-7°; Boiling Point: 80.7°; Flash point: -18°C (-1°F)
Density (g/mL): 0.779; Refractive Index: 1.426


Formula: C₆H₁₂
MW: 84,16 g/mol
Boiling Pt: 81 °C (1013 hPa)
Melting Pt: 7 °C
Density: 0.7785 g/cm³ (20 °C)
Flash Pt: -18 °C
Storage Temperature: ambient
MDL Number: MFCD00003814
CAS Number: 110-82-7
EINECS: 203-806-2
UN: 1145
ADR: 3,II
Merck Index: 13,02752
https://us.vwr.com/stibo/web/std.lang.all/23/06/772306.jpg
Order Now
Specification Test Results
Assay 99.7% min.


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty