Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phthalic anhydride ≥99%

Supplier: BeanTown Chemical


214145-5KG 214145-1KG
BT214145-5KGEA 63.19 USD InStock
BT214145-5KG BT214145-1KG
Phthalic anhydride ≥99%
Phthalic anhydride
CAS: 85-44-9; EC No: 201-607-5; MDL No: MFCD00005918; RTECS: TI3150000
UN No: UN2214; Haz Class: 8; Packing Group: III
Crystalline/Flakes; Molecular Formula: C8H4O3; MW: 148.12
Melting Point: 131-134°; Boiling Point: 284°; Flash point: 152°C (306°F)
Density (g/mL): 1.53
Moisture Sensitive


Formula: C₈H₄O₃
MW: 148,12 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 131 °C
Density: 1.545 g/cm³ (20 °C)
Flash Pt: 151 °C
Storage Temperature: ambient
MDL Number: MFCD00005918
CAS Number: 85-44-9
EINECS: 201-607-5
UN: 2214
ADR: 8,III
Merck Index: 13,07457
Order Now
Specification Test Results
Assay 99%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty