4-Pentylphenol ≥98%
Supplier: BeanTown Chemical
Synonyms:
4-Amylphenol
214110-100G
214110-25G
214110-5G
BT214110-100GEA
923.4
USD
BT214110-100G
BT214110-5G
BT214110-25G
4-Pentylphenol ≥98%
4-Pentylphenol
- RTECS: SM6750000
- Refractive index: 1.515
- Solid
Formula:
C₁₁H₁₆O MW: 164.25 g/mol Melting Pt: 23…25 °C Density: 0.96 g/mL Flash Pt: 100 °C (212 °F) |
MDL Number:
MFCD00020211 CAS Number: 14938-35-3 EINECS: 239-015-4 UN: 2430 ADR: 8,III |
Specification Test Results
Assay | 98% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...