(Diethylamino)sulfur trifluoride ≥95%

Supplier: BeanTown Chemical

Synonyms: DAST

214105-5G 214105-25G 214105-100G
BT214105-25GEA 282.53 USD
BT214105-25G BT214105-100G BT214105-5G
(Diethylamino)sulfur trifluoride ≥95%
(Diethylamino)sulphur trifluoride

  • Moisture Sensitive
  • Liquid

Formula: C₄H₁₀F₃NS
MW: 161.19 g/mol
Boiling Pt: 30…32 °C/3 mmHg
Density: 1.22 g/mL
Flash Pt: 23 °C (73 °F)
MDL Number: MFCD00000363
CAS Number: 38078-09-0
EINECS: 253-771-2
UN: 2920
ADR: 8,I

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR