1H,1H,2H,2H-Perfluorooctyltrichlorosilane ≥97%

Supplier: BeanTown Chemical

Synonyms: Tridecafluoro-1,1,2,2-tetrahydrooctyl-1-trichlorosilane

214030-50G 214030-10G
BT214030-10GEA 98.9 USD
BT214030-10G BT214030-50G
1H,1H,2H,2H-Perfluorooctyltrichlorosilane ≥97%
1H,1H,2H,2H-Perfluorooctyltrichlorosilane

  • Refractive index: 1.352
  • Moisture Sensitive
  • Liquid

Formula: C₈H₄Cl₃F₁₃Si
Boiling Pt: 192 °C
Density: 1.3 g/mL
Flash Pt: 87 °C (189 °F)
MDL Number: MFCD00042363
CAS Number: 78560-45-9
EINECS: 278-947-6
UN: 2987
ADR: 8,II

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR