Propargyl methacrylate ≥98% stabilized

Supplier: BeanTown Chemical

213945-50G 213945-10G
BT213945-10GEA 85.9 USD
BT213945-10G BT213945-50G
Propargyl methacrylate ≥98% stabilized
Propargyl methacrylate

  • Refractive index: 1.445
  • Liquid

Formula: C₇H₈O₂
Boiling Pt: 149…151 °C
Density: 0.986 g/mL
Flash Pt: 48 °C (118 °F)
MDL Number: MFCD00048123
CAS Number: 13861-22-8
EINECS: 237-599-5
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR