Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ω-Bromo-4-cyanoacetophenone ≥97%

Supplier: BeanTown Chemical


213880-25G 213880-5G
BT213880-25GEA 112.56 USD InStock
BT213880-25G BT213880-5G
ω-Bromo-4-cyanoacetophenone ≥97%
ω-Bromo-4-cyanoacetophenone
CAS: 20099-89-2; MDL No: MFCD00052931
UN No: UN2923; Haz Class: 8 (6.1); Packing Group: II
Crystalline/Powder; Molecular Formula: C9H6BrNO; MW: 224.05
Melting Point: 92-96°


Formula: C₉H₆BrNO
MW: 224.06 g/mol
Boiling Pt: 314 °C (1013 hPa)
Melting Pt: 92 to 94 °C
MDL Number: MFCD00052931
CAS Number: 20099-89-2
EINECS: 000-000-0
UN: 2923
ADR: 8,II
Order Now
Specification Test Results
Assay 97%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty