Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Cadmium acetate dihydrate ≥98%

Supplier: BeanTown Chemical

Synonyms: Cadmium di(acetate) dihydrateAcetic acid cadmium salt dihydrate

213790-100G 213790-2KG 213790-500G
BT213790-100GEA 45.29 USD InStock
BT213790-100G BT213790-2KG BT213790-500G
Cadmium acetate dihydrate ≥98%
Cadmium acetate dihydrate
CAS: 5743-04-4; EC No: 208-853-2; MDL No: MFCD00012449
UN No: UN2570; Haz Class: 6.1; Packing Group: III
Powder; Linear Formula: Cd(OOCCH3)2·2H2O; MW: 266.53
Hygroscopic


Formula: (H₃CCOO)₂Cd·2H₂O
MW: 266,53 g/mol
Boiling Pt: 105 to 107 °C (4 torr)
Melting Pt: 256 °C
Density: 2.341 g/cm³ (20 °C)
Storage Temperature: ambient
MDL Number: MFCD00150020
CAS Number: 5743-04-4
EINECS: 208-853-2
UN: 2570
ADR: 6.1,III
Merck Index: 13,01614
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty