3-Picolyl chloride hydrochloride ≥97%
Supplier: BeanTown Chemical
Synonyms:
3-(Chloromethyl)pyridine hydrochloride
, 3-(Chloromethyl)-pyridinium chloride
213785-100G
213785-5G
213785-25G
BT213785-5GEA
27.61
USD
BT213785-5G
BT213785-25G
BT213785-100G
3-Picolyl chloride hydrochloride ≥97%
3-Picolyl chloride hydrochloride
- RTECS: US7000000
- Hygroscopic
- Solid
Formula:
C₆H₇Cl₂N MW: 164.03 g/mol Melting Pt: 137…143 °C |
MDL Number:
MFCD00012818 CAS Number: 6959-48-4 EINECS: 230-150-4 UN: 3261 ADR: 8,III |
Specification Test Results
Assay | 97% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...