Iron(II) bromide, hydrate
Supplier: BeanTown Chemical
|
Danger
|
213730-100G
213730-25G
BT213730-100GEA
207.72
USD
BT213730-100G
BT213730-25G
Iron(II) bromide, hydrate
Iron (II) bromide
- Air Sensitive, Hygroscopic
Formula:
Br₂Fe MW: 215.66 g/mol |
MDL Number:
MFCD00016081 CAS Number: 7789-46-0 EINECS: 232-168-8 UN: 3260 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...