2,3,5-Trifluoropyridine ≥98%

Supplier: BeanTown Chemical

213705-5G 213705-1G 213705-25G
BT213705-1GEA 38.84 USD
BT213705-1G BT213705-25G BT213705-5G
2,3,5-Trifluoropyridine ≥98%
2,3,5-Trifluoropyridine

  • Liquid

Formula: C₅H₂F₃N
MW: 133.07 g/mol
Density: 1.400 g/mL
Flash Pt: 30 °C (86 °F)
MDL Number: MFCD03001162
CAS Number: 76469-41-5
EINECS: 000-000-0
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR