Copper ≥99.9% (trace metals basis), flakes 1-5 µm
Supplier: BeanTown Chemical
213665-100G
213665-500G
BT213665-500GEA
145.37
USD
BT213665-500G
BT213665-100G
Copper ≥99.9% (trace metals basis), flakes 1-5 µm
Copper
- Solid
Formula:
Cu MW: 63.55 g/mol Boiling Pt: 2567 °C Melting Pt: 1083.4 °C Density: 8.94 g/mL |
MDL Number:
MFCD00010965 CAS Number: 7440-50-8 EINECS: 231-159-6 ADR: 4.1,III |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...