1,2,5,6,9,10-Hexabromocyclododecane ≥95%

Supplier: BeanTown Chemical

213585-25G
BT213585-25GEA 22.34 USD
BT213585-25G
1,2,5,6,9,10-Hexabromocyclododecane ≥95%
1,2,5,6,9,10-Hexabromocyclododecane

  • Powder

Formula: C₁₂H₁₈Br₆
MW: 641.7 g/mol
Boiling Pt: >190 °C
Melting Pt: 188…191 °C
CAS Number: 3194-55-6
EINECS: 221-695-9
ADR: 9,III

Order Now

Specification Test Results

Assay 95%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR