Diethylzinc ≥96%

Supplier: BeanTown Chemical

213580-100G
BT213580-100GEA 274.08 USD
BT213580-100G
Diethylzinc ≥96%
Diethylzinc

  • Refractive index: 1.498
  • Air Sensitive, Moisture Sensitive
  • Liquid, packaged in a non-returnable cylinder

Formula: C₄H₁₀Zn
Boiling Pt: 117 °C
Melting Pt: –28 °C
Density: 1.205 g/mL
Flash Pt: –18 °C (–1 °F)
MDL Number: MFCD00009021
CAS Number: 557-20-0
EINECS: 209-161-3
ADR: 4.2 (4.3),I

Order Now

Specification Test Results

Assay 96%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR