Sodium dicyanoaurate ≥99.9% (trace metals basis)
Synonyms:
Gold(I) sodium cyanide
213335-5G
213335-1G
BT213335-1GEA
317.73
USD
BT213335-1G
BT213335-5G
Sodium dicyanoaurate ≥99.9% (trace metals basis)
Sodium dicyanoaurate
- Powder
Formula:
C₂AuN₂Na |
MDL Number:
MFCD00050423 CAS Number: 15280-09-8 EINECS: 239-320-2 UN: 1588 ADR: 6.1,II |
Specification Test Results
Assay | 99.9% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...