Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to system maintenance, custom pricing and product availability will not be shown from 4pm ET Saturday through 8pm Sunday.

Pentachloropyridine ≥98%

Supplier: BeanTown Chemical

Synonyms: 2,3,4,5,6-PentachloropyridinePerchloropyridine

213170-100G 213170-500G 213170-2.5KG
BT213170-100GEA 59.69 USD InStock
BT213170-100G BT213170-500G BT213170-2.5KG
Pentachloropyridine ≥98%
Pentachloropyridine
CAS: 2176-62-7; EC No: 218-535-5; MDL No: MFCD00006230; RTECS: UT7000000
UN No: UN3077; Haz Class: 9; Packing Group: III
Powder; Molecular Formula: C5Cl5N; MW: 251.33
Melting Point: 123-127°


Formula: C₅Cl₅N
MW: 251,33 g/mol
Boiling Pt: 279 to 280 °C (1013 hPa)
Melting Pt: 124 °C
Storage Temperature: ambient
CAS Number: 2176-62-7
EINECS: 218-535-5
UN: 3077
ADR: 9,III
Order Now
Specification Test Results
Assay 98%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty