Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Benzene-[D₆] ≥100% (Isotopic) contains 0.03% TMS

Supplier: BeanTown Chemical

Synonyms: Benzene-[²H₆]

213070-10EACH 213070-2EACH
BT213070-10EAEA 153.17 USD InStock
BT213070-10EA BT213070-2EACH
Benzene-[D₆] ≥100% (Isotopic) contains 0.03% TMS
Benzene-[D6]
CAS: 1076-43-3; EC No: 214-061-8; MDL No: MFCD00003010
UN No: UN1114; Haz Class: 3; Packing Group: II
Liquid, in prescored ampules, 0.75ml/ampule; Molecular Formula: C6D6; MW: 84.15
Melting Point: 6.8°; Boiling Point: 79.1°; Flash point: -11°C (12°F)
Density (g/mL): 0.95; Refractive Index: 1.497
Moisture Sensitive


Formula: C₆D₆
MW: 84,07 g/mol
Boiling Pt: 79 °C (1013 hPa)
Melting Pt: 6.7 °C
Density: 0.95 g/cm³ (20 °C)
Flash Pt: -11 °C (closed cup)
Storage Temperature: ambient
MDL Number: MFCD00003010
CAS Number: 1076-43-3
EINECS: 214-061-8
UN: 1114
ADR: 3,II
Order Now
Specification Test Results
Assay 100%


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty