Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Ammonium perchlorate

Supplier: BeanTown Chemical


213010-250G 213010-1KG
BT213010-250GEA 39.61 USD InStock
BT213010-250G BT213010-1KG
Ammonium perchlorate
Ammonium perchlorate
CAS: 7790-98-9; EC No: 232-235-1; MDL No: MFCD00011421
UN No: UN1442; Haz Class: 5.1; Packing Group: II
Crystalline; Linear Formula: NH4ClO4; MW: 117.49
Density (g/mL): 1.95


Formula: H₄ClNO₄
MW: 117,49 g/mol
Density: 1.95 g/cm³ (20 °C)
MDL Number: MFCD00011421
CAS Number: 7790-98-9
EINECS: 232-235-1
UN: 1442
ADR: 5.1,II
Merck Index: 12,00574
Order Now


 

ORDER

RELATED INFORMATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty