Sodium cyanide ≥98%

Supplier: BeanTown Chemical

212960-1KG 212960-50G 212960-250G
BT212960-250GEA 38.99 USD
BT212960-250G BT212960-50G BT212960-1KG
Sodium cyanide ≥98%
Sodium cyanide

  • RTECS: VZ7525000
  • Hygroscopic
  • Crystalline

Formula: CNNa
MW: 49.01 g/mol
Boiling Pt: 1500 °C
Melting Pt: 564 °C
Density: 1.61 g/mL
MDL Number: MFCD00003523
CAS Number: 143-33-9
EINECS: 205-599-4
UN: 1689
ADR: 6.1,I

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR