2-Methoxypyridine ≥98%

Supplier: BeanTown Chemical

212895-25G 212895-100G 212895-500G
BT212895-100GEA 98.85 USD
BT212895-100G BT212895-500G BT212895-25G
2-Methoxypyridine ≥98%
2-Methoxypyridine

  • Refractive index: 1.503
  • Liquid

Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 142 °C
Density: 1.038 g/mL
Flash Pt: 32 °C (90 °F)
MDL Number: MFCD00006262
CAS Number: 1628-89-3
EINECS: 216-623-8
UN: 3271
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR