Titanium disilicide ≥99.5% (trace metals basis)

Supplier: BeanTown Chemical

Synonyms: Titanium silicide

212585-50G 212585-250G
BT212585-250GEA 364.56 USD
BT212585-250G BT212585-50G
Titanium disilicide ≥99.5% (trace metals basis)
Titanium disilicide

  • -325 Mesh Powder

Formula: Si₂Ti
MW: 104.07 g/mol
Melting Pt: 1760 °C
MDL Number: MFCD00151541
CAS Number: 12039-83-7
EINECS: 234-904-3
ADR: 4.1,III

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR