Calcium iodate ≥98%

Supplier: BeanTown Chemical

212565-100G
BT212565-100GEA 44.01 USD
BT212565-100G
Calcium iodate ≥98%
Calcium iodate

  • Powder

Formula: CaI₂O₆
Melting Pt: 540 °C
Density: 4.520 g/mL
MDL Number: MFCD00036265
CAS Number: 7789-80-2
EINECS: 232-191-3
ADR: 5.1,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR