Cyclopentene ≥97%
Supplier: BeanTown Chemical
212555-500ML
212555-100ML
BT212555-500MLEA
163.04
USD
BT212555-500ML
BT212555-100ML
Cyclopentene ≥97%
Cyclopentene
- RTECS: GY5950000
- Refractive index: 1.421
- Air Sensitive
- Liquid
Formula:
C₅H₈ Boiling Pt: 44…46 °C Melting Pt: –135 °C Density: 0.771 g/mL Flash Pt: –34 °C (–29 °F) |
MDL Number:
MFCD00001394 CAS Number: 142-29-0 EINECS: 205-532-9 UN: 2246 ADR: 3,II |
Specification Test Results
Assay | 97% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...