Perchloric acid ≥70%

Supplier: BeanTown Chemical

211600-250ML 211600-50ML 211600-5ML
BT211600-5MLEA 51.82 USD
BT211600-5ML BT211600-50ML BT211600-250ML
Perchloric acid ≥70%
Perchloric acid

  • RTECS: SC7500000
  • Liquid

Formula: HClO₄
MW: 100.46 g/mol
Boiling Pt: 203 °C
Melting Pt: –112 °C
Density: 1.664 g/mL
Flash Pt: >110 °C (>230 °F)
MDL Number: MFCD00011325
CAS Number: 7601-90-3
EINECS: 231-512-4
UN: 1873
ADR: 5.1,I
https://us.vwr.com/stibo/web/std.lang.all/06/22/16640622.jpg

Order Now

Specification Test Results

Assay 99.995%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR