Indium(I) chloride, anhydrous ≥99.995% (trace metals basis)
Packaged under argon.
211545-500MG
211545-5G
BT211545-5GEA
103.91
USD
BT211545-5G
BT211545-500MG
Indium(I) chloride, anhydrous ≥99.995% (trace metals basis)
Indium (I) chloride
- Moisture Sensitive
- Crystalline
Formula:
InCl MW: 150.27 g/mol Boiling Pt: 608 °C Melting Pt: 225 °C |
MDL Number:
MFCD00148875 CAS Number: 13465-10-6 EINECS: 236-693-3 UN: 3260 ADR: 8,III |
Specification Test Results
Assay | 99.995% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...