Tetramethyltin ≥98%

Supplier: BeanTown Chemical

Synonyms: Tin tetramethylTetramethylstannane

211510-25G 211510-5G
BT211510-25GEA 135.01 USD
BT211510-25G BT211510-5G
Tetramethyltin ≥98%
Tetramethyltin

  • Refractive index: 1.441
  • Liquid

Formula: C₄H₁₂Sn
MW: 178.85 g/mol
Boiling Pt: 74…75 °C
Melting Pt: –54 °C
Density: 1.291 g/mL
Flash Pt: –12 °C (10 °F)
MDL Number: MFCD00008278
CAS Number: 594-27-4
EINECS: 209-833-6
UN: 1992
ADR: 3,II

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR