Dodecacarbonyltriruthenium ≥99%
Supplier: BeanTown Chemical
Synonyms:
Ruthenium carbonyl
, Triruthenium dodecacarbonyl
145915-5G
145915-1G
BT145915-1GEA
127.35
USD
BT145915-1G
BT145915-5G
Dodecacarbonyltriruthenium ≥99%
Dodecacarbonyltriruthenium
- Crystalline
Formula:
C₁₂O₁₂Ru₃ MW: 639.33 g/mol |
EINECS:
239-287-4 UN: 3466 ADR: 6.1,III |
Specification Test Results
Assay | 99% |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...