BEAN145445

145445-100G 145445-25G 145445-500G
BT145445-25GEA 65.32 USD
BT145445-25G BT145445-500G BT145445-100G
BEAN145445
CAS: 14456-34-9; EC No: 237-345-3; MDL No: MFCD00150417; RTECS: MG4800000
UN No: UN3260; Haz Class: 8; Packing Group: III
Powder; Linear Formula: HfOCl2·8H2O; MW: 409.52
Hygroscopic
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR