di-Lithium oxalate ≥99%

Supplier: BeanTown Chemical

Synonyms: Lithium oxalateDilithium oxalateLithiumoxalate

145315-1KG 145315-250G 145315-50G
BT145315-50GEA 36.77 USD
BT145315-50G BT145315-1KG BT145315-250G
di-Lithium oxalate ≥99%
di-Lithium oxalate

  • Powder

Formula: C₂Li₂O₄
MW: 101.9 g/mol
MDL Number: MFCD00040596
CAS Number: 553-91-3
EINECS: 209-054-1
UN: 3288
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR