2-Ethyl-2-oxazoline ≥99%

Supplier: BeanTown Chemical

144650-100G
BT144650-100GEA 35.66 USD
BT144650-100G
2-Ethyl-2-oxazoline ≥99%
2-Ethyl-2-oxazoline

  • Refractive index: 1.437
  • Air Sensitive
  • Liquid

Formula: C₅H₉NO
Boiling Pt: 128 °C
Melting Pt: –62 °C
Density: 0.982 g/mL
Flash Pt: 29 °C (84 °F)
MDL Number: MFCD00005302
CAS Number: 10431-98-8
EINECS: 233-912-4
ADR: 8 (3),II

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR