BEAN144310

144310-500ML 144310-100ML
BT144310-100MLEA 39.05 USD
BT144310-100ML BT144310-500ML
BEAN144310
CAS: 7803-49-8; EC No: 232-259-2; MDL No: MFCD00044522; RTECS: NC2975000
UN No: UN3266; Haz Class: 8; Packing Group: III
Liquid; Linear Formula: NH2OH; MW: 33.03
Boiling Point: 107-112°
Density (g/mL): 1.078
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR