Titanium(III) chloride 20% (w/w) in hydrochloric acid 3%

Supplier: BeanTown Chemical

144105-5KG 144105-1KG 144105-250G
BT144105-250GEA 56.11 USD
BT144105-250G BT144105-1KG BT144105-5KG
Titanium(III) chloride 20% (w/w) in hydrochloric acid 3%
Titanium (III) chloride

  • Air Sensitive
  • Liquid

Formula: Cl₃Ti
MW: 154.24 g/mol
Density: 1.192 g/mL
MDL Number: MFCD00011266
CAS Number: 7705-07-9
EINECS: 231-728-9
UN: 3264
ADR: 4.2,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR