N,N-Dimethylglycine ethyl ester ≥98%

Supplier: BeanTown Chemical

Synonyms: Di-Me-Gly-OEt

143165-25G
BT143165-25GEA 63.67 USD
BT143165-25G
N,N-Dimethylglycine ethyl ester ≥98%
N,N-Dimethylglycine ethyl ester

  • RTECS: MB9990000
  • Refractive index: 1.413
  • Air Sensitive, Hygroscopic, Moisture Sensitive
  • Liquid

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
Boiling Pt: 150…151 °C
Density: 0.928 g/mL
Flash Pt: 32 °C (90 °F)
MDL Number: MFCD00009175
CAS Number: 33229-89-9
EINECS: 251-411-9
ADR: 3,III

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR