Barium acetate 99.0-102.0% ACS

Supplier: BeanTown Chemical

Synonyms: Barium di(acetate)Acetic acid barium salt

142020-250G 142020-5KG 142020-1KG
BT142020-5KGEA 547.37 USD
BT142020-5KG BT142020-250G BT142020-1KG
Barium acetate 99.0-102.0% ACS
Barium acetate

  • Crystalline

Formula: (H₃CCOO)₂Ba
MW: 255.42 g/mol
MDL Number: MFCD00012447
CAS Number: 543-80-6
EINECS: 208-849-0
UN: 1564
ADR: 6.1,III

Order Now

Specification Test Results

Assay 99.0-102.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR