Indene ≥90% stabilized, Technical Grade
Supplier: BeanTown Chemical
Synonyms:
1H-Indene
141765-1L
141765-50ML
141765-250ML
BT141765-250MLEA
64.81
USD
BT141765-250ML
BT141765-50ML
BT141765-1L
Indene ≥90% stabilized, Technical Grade
Indene
- RTECS: NK8225000
- Refractive index: 1.595
- Liquid
Formula:
C₉H₈ MW: 116.16 g/mol Boiling Pt: 181…182 °C Melting Pt: –5 °C Density: 0.996 g/mL Flash Pt: 58 °C (136 °F) |
MDL Number:
MFCD00003777 CAS Number: 95-13-6 EINECS: 202-393-6 UN: 3295 ADR: 3,III |
Specification Test Results
Assay | 90% |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...